Stampa…

3-Fluoro-4-nitrobenzaldehyde ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

F1130-1G F1130-5G
TCIAF1130-1GEA 58 EUR
TCIAF1130-1G TCIAF1130-5G
3-Fluoro-4-nitrobenzaldehyde ≥97.0% (by GC)
3-Fluoro-4-nitrobenzaldehyde
Formula: C₇H₄NO₃F
Peso molecolare: 169,1100 g/mol
MDL Number: MFCD00968940
CAS Number: 160538-51-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=757b642817fe8a4643baab2c15168b61d2c00b57&vid=757b642817fe8a4643baab2c15168b61d2c00b57&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 53.0 to 57.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR