[4-Fluoro-3-(trifluoromethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium p-Toluenesulphonate ≥97.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
[4-Fluoro-3-(trifluoromethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium Tosylate
F1111-200MG
F1111-1G
TCIAF1111-200MGEA
74
EUR
TCIAF1111-200MG
TCIAF1111-1G
[4-Fluoro-3-(trifluoromethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium p-Toluenesulphonate ≥97.0% (by HPLC)
[4-Fluoro-3-(trifluoromethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium p-Toluenesulphonate
Formula:
C₂₃H₂₁F₄IO₆S Peso molecolare: 628,37 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD30475655 CAS Number: 1868173-29-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 % (total of Ion-Pair) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...