Stampa…

2-Formyl-4-methoxyphenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI
Attenzione

F1108-1G F1108-5G
TCIAF1108-1GEA 65 EUR
TCIAF1108-1G TCIAF1108-5G
2-Formyl-4-methoxyphenylboronic acid (contains varying amounts of Anhydride)
2-Formyl-4-methoxyphenylboronic acid
Formula: C₈H₉BO₄
Punto di fusione: 161°C
MDL Number: MFCD01319043
CAS Number: 139962-95-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 98.0 to 111.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR