Stampa…

trans-4-Fluoro-4'-(4-n-propylcyclohexyl)biphenyl ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 4-Fluoro-4'-(trans-4-propylcyclohexyl)biphenyl, 4-Fluoro-4'-(trans-4-propylcyclohexyl)-1,1'-biphenyl

F1103-1G F1103-5G
TCIAF1103-1GEA 25 EUR
TCIAF1103-1G TCIAF1103-5G
trans-4-Fluoro-4'-(4-n-propylcyclohexyl)biphenyl ≥98.0% (by GC)
trans-4-Fluoro-4'-(4-n-propylcyclohexyl)biphenyl
Formula: C₂₁H₂₅F
Peso molecolare: 296,429 g/mol
MDL Number: MFCD11053438
CAS Number: 87260-24-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcd247ab55c29e2c5aa5e1afade1b0b08899c1ab&vid=dcd247ab55c29e2c5aa5e1afade1b0b08899c1ab&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 104 to 160 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR