Stampa…

3-Fluoro-2-formylphenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI
Pericolo

Synonyms: (3-Fluoro-2-formylphenyl)boronic acid, 3-Fluoro-2-formylbenzeneboronic acid

F1089-1G F1089-5G
TCIAF1089-1GEA 37 EUR
TCIAF1089-1G TCIAF1089-5G
3-Fluoro-2-formylphenylboronic acid (contains varying amounts of Anhydride)
3-Fluoro-2-formylphenylboronic acid
Formula: C₇H₆BFO₃
Punto di fusione: 129°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD10697421
CAS Number: 871126-15-7
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f00214a8b704cf004bdcb3c333ba91a9eb19042c&vid=f00214a8b704cf004bdcb3c333ba91a9eb19042c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 113.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR