Stampa…

3-Formyl-2-methoxyphenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI
Attenzione

F1088-1G F1088-5G
TCIAF1088-1GEA 66 EUR
TCIAF1088-1G TCIAF1088-5G
3-Formyl-2-methoxyphenylboronic acid (contains varying amounts of Anhydride)
3-Formyl-2-methoxyphenylboronic acid
Formula: C₈H₉BO₄
Peso molecolare: 179.9700 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD08689486
CAS Number: 480424-49-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf7b32dfc20428a2cad52a7b7b7b7e774da73bc2&vid=cf7b32dfc20428a2cad52a7b7b7b7e774da73bc2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR