Stampa…

6-Fluorochromane-2-carboxylic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

F1086-5G F1086-25G
TCIAF1086-5GEA 85 EUR
TCIAF1086-5G TCIAF1086-25G
6-Fluorochromane-2-carboxylic acid ≥98.0% (by GC, titration analysis)
6-Fluorochromane-2-carboxylic acid
Formula: C₁₀H₉FO₃
MDL Number: MFCD07778357
CAS Number: 99199-60-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff211675a0a0181c5f19c5d0205b3a30de027c60&vid=ff211675a0a0181c5f19c5d0205b3a30de027c60&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 124.0 to 128.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR