Stampa…

5-Fluororesorcinol ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Fluorobenzene-1,3-diol

F1081-200MG F1081-1G
TCIAF1081-200MGEA 189 EUR
TCIAF1081-200MG TCIAF1081-1G
5-Fluororesorcinol ≥98.0% (by GC, titration analysis)
5-Fluororesorcinol
Formula: C₆H₅FO₂
Peso molecolare: 128,10 g/mol
MDL Number: MFCD08457182
CAS Number: 75996-29-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 136.0 to 140.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR