Stampa…

3-Fluoro-5-methylphenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (3-Fluoro-5-methylphenyl)boronic acid, 3-Fluoro-5-methylbenzeneboronic acid

F1068-1G F1068-5G
TCIAF1068-1GEA 91 EUR
TCIAF1068-1G TCIAF1068-5G
3-Fluoro-5-methylphenylboronic acid (contains varying amounts of Anhydride)
3-Fluoro-5-methylphenylboronic acid
Formula: C₇H₈BFO₂
Peso molecolare: 153,95 g/mol
MDL Number: MFCD07363785
CAS Number: 850593-06-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4be069eb3814ab1620c28212affbd7c2bf77c44b&vid=4be069eb3814ab1620c28212affbd7c2bf77c44b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 98.0 to 114.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR