1-(4-Fluorophenyl)-1H-pyrrole-2,5-dione ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
N-(4-Fluorophenyl)maleimide
F1054-200MG
F1054-1G
TCIAF1054-200MGEA
33
EUR
TCIAF1054-200MG
TCIAF1054-1G
1-(4-Fluorophenyl)-1H-pyrrole-2,5-dione ≥98.0% (by GC)
1-(4-Fluorophenyl)-1H-pyrrole-2,5-dione
Formula:
C₁₀H₆FNO₂ Peso molecolare: 191,16 g/mol |
MDL Number:
MFCD00033653 CAS Number: 6633-22-3 |
Risultati del test di specificazione
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 154.0 to 158.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...