Stampa…

5-Fluoro-3-methylindole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

F1053-200MG F1053-1G
TCIAF1053-200MGEA 36 EUR
TCIAF1053-200MG TCIAF1053-1G
5-Fluoro-3-methylindole ≥98.0% (by GC)
5-Fluoro-3-methylindole
Formula: C₉H₈FN
Peso molecolare: 149,17 g/mol
MDL Number: MFCD03095377
CAS Number: 392-13-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 83.0 to 87.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR