Stampa…

2-Fluoro-4-methoxyphenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Fluoro-4-methoxybenzeneboronic acid

F1048-1G F1048-5G
TCIAF1048-1GEA 104 EUR
TCIAF1048-1G TCIAF1048-5G
2-Fluoro-4-methoxyphenylboronic acid (contains varying amounts of Anhydride)
2-Fluoro-4-methoxyphenylboronic acid
Formula: C₇H₈BFO₃
Peso molecolare: 169,95 g/mol
MDL Number: MFCD04039323
CAS Number: 162101-31-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f347e7e51acc6de85b4c996f7a426e05d4ac2047&vid=f347e7e51acc6de85b4c996f7a426e05d4ac2047&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR