Fmoc-Lys(Mtt)-OH ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
Synonyms:
Nα-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-(4-methyltrityl)-L-lysine, Nα-Fmoc-Nepsilon-(4-methyltrityl)-L-lysine
F1043-1G
F1043-5G
TCIAF1043-1GEA
39
EUR
TCIAF1043-1G
TCIAF1043-5G
Fmoc-Lys(Mtt)-OH ≥98.0% (by HPLC, total nitrogen)
Fmoc-Lys(Mtt)-OH
Formula:
C₄₁H₄₀N₂O₄ Peso molecolare: 624,781 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00237166 CAS Number: 167393-62-6 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Specific rotation [a]20/D | -9.0 to -11.0 deg (C=1,DMF) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...