Stampa…

2-Fluoro-4-iodo-5-methylpyridine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Fluoro-4-iodo-5-picoline

F1035-1G F1035-5G
TCIAF1035-1GEA 23 EUR
TCIAF1035-1G TCIAF1035-5G
2-Fluoro-4-iodo-5-methylpyridine ≥98.0% (by GC)
2-Fluoro-4-iodo-5-methylpyridine
Formula: C₆H₅FIN
Peso molecolare: 237,02 g/mol
MDL Number: MFCD03095297
CAS Number: 153034-94-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 42.0 to 46.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR