Stampa…

7-Fluoroquinazolin-4(3H)-one ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 7-Fluoro-4-hydroxyquinazoline, 7-Fluoro-4-quinazolinone, 7-Fluoro-4-quinazolinol

F1029-1G F1029-5G
TCIAF1029-1GEA 22 EUR
TCIAF1029-1G TCIAF1029-5G
7-Fluoroquinazolin-4(3H)-one ≥98.0% (by GC, titration analysis)
7-Fluoroquinazolin-4(3H)-one
Formula: C₈H₅FN₂O
Peso molecolare: 164,1400 g/mol
MDL Number: MFCD05664874
CAS Number: 16499-57-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Red powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 246.0 to 250.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR