Stampa…

Fmoc-L-homophenylalanine ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: Fmoc-HoPhe-OH, (S)-2-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]-4-phenylbutyric acid, Fmoc-Homophe-OH, N-Fmoc-L-homophenylalanine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-homophenylalanine, (S)-2-(Fmoc-amino)-4-phenylbutyric acid

F1025-1G F1025-5G
TCIAF1025-1GEA 79 EUR
TCIAF1025-1G TCIAF1025-5G
Fmoc-L-homophenylalanine ≥98.0% (by HPLC)
Fmoc-L-homophenylalanine
Formula: C₂₅H₂₃NO₄
Peso molecolare: 401,462 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00077046
CAS Number: 132684-59-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 95.0 %
Melting point 141.0 to 145.0 deg-C
Specific rotation [a]20/D -6.0 to -9.0 deg (C=1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR