3-(4-Fluorophenyl)-5-methylisoxazole-4-carboxylic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-(4-Fluorophenyl)-5-methylisoxazol-4-carboxylic acid
F1023-1G
F1023-5G
TCIAF1023-1GEA
41
EUR
TCIAF1023-1G
TCIAF1023-5G
3-(4-Fluorophenyl)-5-methylisoxazole-4-carboxylic acid ≥98.0% (by GC, titration analysis)
3-(4-Fluorophenyl)-5-methylisoxazole-4-carboxylic acid
Formula:
C₁₁H₈FNO₃ Peso molecolare: 221,19 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD03407356 CAS Number: 1736-21-6 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 200.0 to 204.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...