3-Fluoro-5-hydroxypyridine ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₅H₄FNO Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD03095199 CAS Number: 209328-55-2 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 162.0 to 166.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...