3-Fluoro-4-(trifluoromethyl)benzonitrile ≥99.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
4-Cyano-2-fluorobenzotrifluoride, 3-Fluoro-4-trifluoromethylbenzonitrile
F1019-1G
F1019-5G
TCIAF1019-1GEA
18
EUR
TCIAF1019-1G
TCIAF1019-5G
3-Fluoro-4-(trifluoromethyl)benzonitrile ≥99.0% (by GC)
3-Fluoro-4-(trifluoromethyl)benzonitrile
Formula:
C₈H₃F₄N Peso molecolare: 189,11 g/mol |
MDL Number:
MFCD00236280 CAS Number: 231953-38-1 UN: 3439 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 46.0 to 50.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...