N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(2-furyl)-L-alanine ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
Fmoc-beta-(2-furyl)-Ala-OH, Fmoc-3-(2-Furyl)-Ala-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(furan-2-yl)propanoic acid, N-Fmoc-3-(2-furyl)-L-alanine
F1013-200MG
F1013-1G
TCIAF1013-200MGEA
61
EUR
TCIAF1013-200MG
TCIAF1013-1G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(2-furyl)-L-alanine ≥98.0% (by titrimetric analysis)
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(2-furyl)-L-alanine
Formula:
C₂₂H₁₉NO₅ Peso molecolare: 377,40 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD01311747 CAS Number: 159611-02-6 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 137.0 to 141.0 deg-C |
Specific rotation [a]20/D | -33.0 to -37.0 deg (C=1,DMF) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...