4-Fluoro-2-methoxyaniline ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
F1003-1G
F1003-5G
TCIAF1003-1GEA
106
EUR
TCIAF1003-1G
TCIAF1003-5G
4-Fluoro-2-methoxyaniline ≥98.0% (by GC, titration analysis)
4-Fluoro-2-methoxyaniline
Formula:
C₇H₈FNO Peso molecolare: 141,14 g/mol Punto di ebollizione: 215°C |
MDL Number:
MFCD00077536 CAS Number: 450-91-9 UN: 2810 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...