Stampa…

Ferrocenecarbaldehyde ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: Ferrocenecarboxaldehyde

F0998-5G F0998-25G
TCIAF0998-5GEA 66 EUR
TCIAF0998-5G TCIAF0998-25G
Ferrocenecarbaldehyde ≥97.0% (by GC, titration analysis)
Ferrocenecarbaldehyde
Formula: C₁₁H₁₀FeO
Peso molecolare: 214,04 g/mol
MDL Number: MFCD00001429
CAS Number: 12093-10-6
EINECS: 235-158-1

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 97.0 %
Purity (Chelometric Titration) min. 97.0 %
Melting point 114.0 to 121.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR