6-Fluorosalicylaldehyde ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Fluoro-6-hydroxybenzaldehyde
F0988-1G
F0988-5G
TCIAF0988-1GEA
110
EUR
TCIAF0988-1G
TCIAF0988-5G
6-Fluorosalicylaldehyde ≥98.0% (by GC, titration analysis)
6-Fluorosalicylaldehyde
Formula:
C₇H₅FO₂ Peso molecolare: 140,11 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD01090996 CAS Number: 38226-10-7 |
Risultati del test di specificazione
Appearance | White to Yellow to Orange powder to lump |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 37.0 to 41.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...