Stampa…

4-Fluorobenzoylacetonitrile ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-(4-Fluorophenyl)-3-oxopropanenitrile

F0987-1G F0987-5G
TCIAF0987-1GEA 47 EUR
TCIAF0987-1G TCIAF0987-5G
4-Fluorobenzoylacetonitrile ≥98.0% (by GC)
4-Fluorobenzoylacetonitrile
Formula: C₉H₆FNO
Peso molecolare: 163,15 g/mol
MDL Number: MFCD00662062
CAS Number: 4640-67-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 83.0 to 87.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR