4-Fluoro-m-toluonitrile ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
F0986-5G
F0986-25G
TCIAF0986-5GEA
48
EUR
TCIAF0986-5G
TCIAF0986-25G
4-Fluoro-m-toluonitrile ≥98.0% (by GC)
4-Fluoro-m-toluonitrile
Formula:
C₈H₆FN Peso molecolare: 135,14 g/mol |
MDL Number:
MFCD03788510 CAS Number: 185147-08-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 54.0 to 58.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...