Stampa…

2-Fluoro-6-idrossibenzonitrile ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

F0985-1G F0985-5G
TCIAF0985-1GEA 25 EUR
TCIAF0985-1G TCIAF0985-5G
2-Fluoro-6-idrossibenzonitrile ≥98.0% (by GC, titration analysis)
2-Fluoro-6-idrossibenzonitrile
Formula: C₇H₄FNO
Peso molecolare: 137,11 g/mol
MDL Number: MFCD03428592
CAS Number: 140675-43-0
UN: 3439
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 155.0 to 159.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR