Stampa…

5-Fluoro-2-hydroxybenzeneboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Fluoro-2-hydroxyphenylboronic acid, (5-Fluoro-2-hydroxyphenyl)boronic acid, 5-Fluoro-2-hydroxybenzeneboronic acid

F0977-1G F0977-5G
TCIAF0977-1GEA 64 EUR
TCIAF0977-1G TCIAF0977-5G
5-Fluoro-2-hydroxybenzeneboronic acid (contains varying amounts of Anhydride)
5-Fluoro-2-hydroxybenzeneboronic acid
Formula: C₆H₆BFO₃
Peso molecolare: 155,92 g/mol
MDL Number: MFCD03095357
CAS Number: 259209-20-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 114.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR