Stampa…

Formamidine hydroiodide, low water content ≥98.0% (by total nitrogen and titration analysis)

Fornitore: TCI EUROPE

Synonyms: FAI, Formamidinium iodide

F0974-1G F0974-5G F0974-25G
TCIAF0974-1GEA 63 EUR
TCIAF0974-1G TCIAF0974-5G TCIAF0974-25G
Formamidine hydroiodide, low water content ≥98.0% (by total nitrogen and titration analysis)
Formamidine hydroiodide
Formula: CH₄N₂·HI
Peso molecolare: 171,97 g/mol
MDL Number: MFCD28369273
CAS Number: 879643-71-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a67dd44ecd31d29488517c33c553ec51e098536&vid=3a67dd44ecd31d29488517c33c553ec51e098536&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystaline
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
water max. 100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR