Stampa…

2-Fluoro-6-methoxybenzonitrile ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

F0969-5G F0969-25G
TCIAF0969-5GEA 38 EUR
TCIAF0969-5G TCIAF0969-25G
2-Fluoro-6-methoxybenzonitrile ≥98.0% (by GC)
2-Fluoro-6-methoxybenzonitrile
Formula: C₈H₆FNO
Peso molecolare: 151,14 g/mol
MDL Number: MFCD00042291
CAS Number: 94088-46-7
EINECS: 302-047-5
UN: 3439
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 102.0 to 106.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR