Stampa…

Fenhexamid ≥98.0% (by HPLC)

Fornitore: TCI

F0967-200MG F0967-1G
TCIAF0967-200MGEA 97 EUR
TCIAF0967-200MG TCIAF0967-1G
Fenhexamid ≥98.0% (by HPLC)
Fenhexamid
Formula: C₁₄H₁₇Cl₂NO₂
Punto di ebollizione: 320°C
MDL Number: MFCD03095700
CAS Number: 126833-17-8
EINECS: 422-530-5
UN: 3077
ADR: 9,III
Merck Index: 14,03974

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 97.0 %
Melting point 151.0 to 155.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR