Flumazenil ≥99.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Ethyl 8-fluoro-5-methyl-6-oxo-5,6-dihydro-4H-benzo[f]imidazo[1,5-a][1,4]diazepine-3-carboxylate
F0958-25MG
F0958-100MG
TCIAF0958-25MGEA
46
EUR
TCIAF0958-25MG
TCIAF0958-100MG
Flumazenil ≥99.0% (by GC)
Flumazenil
Formula:
C₁₅H₁₄FN₃O₃ Peso molecolare: 303,29 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00242764 CAS Number: 78755-81-4 Merck Index: 14,04135 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 198.0 to 202.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...