Stampa…

4-Fluoro-3-nitrobenzaldehyde ≥95.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

F0956-1G F0956-5G
TCIAF0956-1GEA 23 EUR
TCIAF0956-1G TCIAF0956-5G
4-Fluoro-3-nitrobenzaldehyde ≥95.0% (by GC)
4-Fluoro-3-nitrobenzaldehyde
Formula: C₇H₄FNO₃
Peso molecolare: 169,11 g/mol
MDL Number: MFCD01861388
CAS Number: 42564-51-2

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 43.0 to 47.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR