Stampa…

6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Paliperidone Related Compound B

F0944-1G F0944-5G
TCIAF0944-1GEA 19 EUR
TCIAF0944-1G TCIAF0944-5G
6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride ≥98.0% (by HPLC, titration analysis)
6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride
Formula: C₁₂H₁₄ClFN₂O
Peso molecolare: 256,7000 g/mol
MDL Number: MFCD06658529
CAS Number: 84163-13-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR