Stampa…

9-Fluorenylmethyl pentafluorophenyl carbonate ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: Carbonic Acid 9-Fluorenylmethyl Pentafluorophenyl Ester, Fmoc-OPfp, 9-Fluorenylmethyl Pentafluorophenyl Carbonate

F0936-1G F0936-5G
TCIAF0936-1GEA 28 EUR
TCIAF0936-1G TCIAF0936-5G
9-Fluorenylmethyl pentafluorophenyl carbonate ≥98.0% (by HPLC)
9-Fluorenylmethyl pentafluorophenyl carbonate
Formula: C₂₁H₁₁F₅O₃
Peso molecolare: 406,31 g/mol
MDL Number: MFCD00013261
CAS Number: 88744-04-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 83.0 to 87.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR