Stampa…

2-Fluorophenethylamine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 1-Amino-2-(2-fluorophenyl)ethane, 2-Fluorophenethylamine, 2-(2-Fluorophenyl)ethylamine

F0933-5G F0933-25G
TCIAF0933-5GEA 50 EUR
TCIAF0933-5G TCIAF0933-25G
2-Fluorophenethylamine ≥98.0% (by GC, titration analysis)
2-Fluorophenethylamine
Formula: C₈H₁₀FN
Peso molecolare: 139,17 g/mol
Flash Pt: 77°C
MDL Number: MFCD00075502
CAS Number: 52721-69-4
UN: 2735
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR