Fmoc-L-4-pyridylalanine ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
N-Fmoc-3-(4-pyridyl)-L-alanine, Fmoc-3-(4-pyridyl)-Ala-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(4-pyridyl)-L-alanine, Fmoc-4-Pal-OH
F0927-1G
F0927-5G
TCIAF0927-1GEA
97
EUR
TCIAF0927-1G
TCIAF0927-5G
Fmoc-L-4-pyridylalanine ≥98.0% (by HPLC, titration analysis)
Fmoc-L-4-pyridylalanine
Formula:
C₂₃H₂₀N₂O₄ Peso molecolare: 388,43 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00672566 CAS Number: 169555-95-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...