Stampa…

4-[[(9H-Fluoren-9-ylmethoxy)carbonyl]aminomethyl]benzoic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI

Synonyms: Fmoc-4-Amb-OH, 4-(Fmoc-aminomethyl)benzoic Acid

F0925-5G F0925-1G
TCIAF0925-5GEA 188 EUR
TCIAF0925-5G TCIAF0925-1G
4-[[(9H-Fluoren-9-ylmethoxy)carbonyl]aminomethyl]benzoic acid ≥98.0% (by HPLC, titration analysis)
4-[[(9H-Fluoren-9-ylmethoxy)carbonyl]aminomethyl]benzoic acid
Formula: C₂₃H₁₉NO₄
Peso molecolare: 373.41 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD01074693
CAS Number: 164470-64-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR