Febantel
Fornitore: TCI EUROPE
Synonyms:
N-(((2-(2-MEthoxyacetamido)-4-(phenylsulphanyl)phenyl)amino)((methoxycarbonyl)imino)methyl)carbamate
F0922-1G
F0922-5G
TCIAF0922-1GEA
58
EUR
TCIAF0922-1G
TCIAF0922-5G
Febantel
Febantel
Formula:
C₂₀H₂₂N₄O₆S Peso molecolare: 446,48 g/mol Punto di fusione: 130°C |
MDL Number:
MFCD01738527 CAS Number: 58306-30-2 EINECS: 261-205-0 Merck Index: 14,03944 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 125.0 to 129.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...