(±)-4-Fluoromandelic acid ≥96.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
F0907-5G
F0907-25G
TCIAF0907-5GEA
81
EUR
TCIAF0907-5G
TCIAF0907-25G
(±)-4-Fluoromandelic acid ≥96.0% (by GC, titration analysis)
(±)-4-Fluoromandelic acid
Formula:
C₈H₇FO₃ |
MDL Number:
MFCD00044402 CAS Number: 395-33-5 EINECS: 206-899-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Neutralization titration) | min. 96.0 % |
Melting point | 135.0 to 139.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...