4-Fluoro-N-(4-fluorobenzylidene)aniline ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
F0892-1G
F0892-5G
TCIAF0892-1GEA
32
EUR
TCIAF0892-1G
TCIAF0892-5G
4-Fluoro-N-(4-fluorobenzylidene)aniline ≥98.0% (by GC, titration analysis)
4-Fluoro-N-(4-fluorobenzylidene)aniline
Formula:
C₁₃H₉F₂N Peso molecolare: 217,22 g/mol |
MDL Number:
MFCD00017950 CAS Number: 39769-09-0 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 63.0 to 67.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...