Fosfomycin disodium salt ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
Formula:
C₃H₅Na₂O₄P Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00056853 CAS Number: 26016-99-9 EINECS: 247-409-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) (CAD) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | -3.5 to -5.5 deg (C=5, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...