Stampa…

2-Fluoro-3-hydroxypyridine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Fluoropyridin-3-ol, 2-Fluoro-3-pyridinol

F0886-1G F0886-5G
TCIAF0886-1GEA 53 EUR
TCIAF0886-1G TCIAF0886-5G
2-Fluoro-3-hydroxypyridine ≥98.0% (by GC, titration analysis)
2-Fluoro-3-hydroxypyridine
Formula: C₅H₄FNO
Peso molecolare: 113,09 g/mol
MDL Number: MFCD04112569
CAS Number: 174669-74-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 126.0 to 131.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR