2-Fluoro-p-toluonitrile ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
F0884-1G
F0884-5G
TCIAF0884-1GEA
45
EUR
TCIAF0884-1G
TCIAF0884-5G
2-Fluoro-p-toluonitrile ≥98.0% (by GC)
2-Fluoro-p-toluonitrile
Formula:
C₈H₆FN Peso molecolare: 135,14 g/mol |
MDL Number:
MFCD03094311 CAS Number: 85070-67-3 UN: 3439 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 51.0 to 55.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...