Stampa…

(3S,4R)-4-(4-Fluorophenyl)-3-hydroxymethyl-1-methylpiperidine ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Pericolo

Synonyms: Paroxol

F0878-5G F0878-25G
TCIAF0878-5GEA 45 EUR
TCIAF0878-5G TCIAF0878-25G
(3S,4R)-4-(4-Fluorophenyl)-3-hydroxymethyl-1-methylpiperidine ≥98.0% (by GC, titration analysis)
(3S,4R)-4-(4-Fluorophenyl)-3-hydroxymethyl-1-methylpiperidine
Formula: C₁₃H₁₈FNO
MDL Number: MFCD06658161
CAS Number: 105812-81-5
EINECS: 406-030-4
UN: 3077
ADR: 9,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 98.0 to 102.0 deg-C
Specific rotation [a]20/D -36.0 to -39.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR