1-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]cyclopentanecarboxylic acid ≥95.0% (by HPLC, titration analysis)
Fornitore: TCI
Formula:
C₂₁H₂₁NO₄ Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD01074696 CAS Number: 117322-30-2 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...