SB203580 ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
SB 203580, 4-(5-(4-Fluorophenyl)-2-(4-(methylsulphinyl)phenyl)-1H-imidazol-4-yl)pyridine
F0864-25MG
F0864-100MG
TCIAF0864-25MGEA
54
EUR
TCIAF0864-25MG
TCIAF0864-100MG
SB203580 ≥98.0% (by HPLC)
SB203580
Formula:
C₂₁H₁₆FN₃OS Peso molecolare: 377,4 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00922198 CAS Number: 152121-47-6 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 243.0 to 247.0 deg-C |
Elemental analysis (Nitrogen) | 10.60 to 11.60 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...