Stampa…

5-Fluoro-3-pyridineboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Fluoropyridine-3-boronic acid, (5-Fluoropyridin-3-yl)boronic acid

F0859-1G F0859-5G
TCIAF0859-1GEA 71 EUR
TCIAF0859-1G TCIAF0859-5G
5-Fluoro-3-pyridineboronic acid (contains varying amounts of Anhydride)
5-Fluoro-3-pyridineboronic acid
Formula: C₅H₅BFNO₂
Peso molecolare: 140,91 g/mol
MDL Number: MFCD07368243
CAS Number: 872041-86-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 115.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR