N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
Fmoc-Cha-OH, N-Fmoc-3-cyclohexyl-L-alanine
F0857-5G
F0857-1G
TCIAF0857-5GEA
96
EUR
TCIAF0857-5G
TCIAF0857-1G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine ≥98.0% (by HPLC, titration analysis)
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine
Formula:
C₂₄H₂₇NO₄ Peso molecolare: 393,48 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00065614 CAS Number: 135673-97-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | -21.0 to -26.0 deg (C=1,DMF) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...