1-(4-Fluorophenyl)-2-thiourea ≥97.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
N-(4-Fluorophenyl)thiourea, 4-Fluorophenylthiourea
F0836-5G
F0836-25G
TCIAF0836-5GEA
41
EUR
TCIAF0836-5G
TCIAF0836-25G
1-(4-Fluorophenyl)-2-thiourea ≥97.0% (by HPLC, total nitrogen)
1-(4-Fluorophenyl)-2-thiourea
Formula:
(C₆H₄F)NHC(S)NH₂ Peso molecolare: 170,21 g/mol |
MDL Number:
MFCD00041180 CAS Number: 459-05-2 UN: 2811 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 158.0 to 162.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...