Stampa…

6-Fluoro-4-chromanone ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 6-Fluorochroman-4-one

F0831-1G F0831-5G
TCIAF0831-1GEA 27 EUR
TCIAF0831-1G TCIAF0831-5G
6-Fluoro-4-chromanone ≥98.0% (by GC)
6-Fluoro-4-chromanone
Formula: C₉H₇FO₂
Peso molecolare: 166,15 g/mol
MDL Number: MFCD00038654
CAS Number: 66892-34-0
EINECS: 266-511-8

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 114.0 to 118.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR